missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIM1, His Tag Recombinant Protein

Product Code. 15448443
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15448443

Brand: Invitrogen™ PR5298C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11309
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.05% Triton X-100, 0.5 mM EDTA, 2 mM DTT, 50% glycerol, 500 mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 5292
Molecular Weight (g/mol) 40.7 kDa
Name Human PIM1, His Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Gene Alias non-specific serine/threonine protein kinase; Oncogene PIM1; PIM; Pim1; Pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); Pim-1 proto-oncogene, serine/threonine kinase; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; serine/threonine-protein kinase pim-1; serine-threonine kinase PIM-1
Common Name PIM1
KinaseFamily PIM Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.