missing translation for 'onlineSavingsMsg'
Learn More

PIM1 Rabbit anti-Human, Mouse, Clone: S08-8D5, Novus Biologicals™

Product Code. 18377514 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377514 100 μg 100µL
18396605 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18377514 Supplier Novus Biologicals Supplier No. NBP314963100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PIM1 Monoclonal antibody specifically detects PIM1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIM1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S08-8D5
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:20-1:50, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias EC 2.7.11.1, Oncogene PIM1, PIM, pim-1 kinase 44 kDa isoform, pim-1 oncogene, pim-1 oncogene (proviral integration site 1), proto-oncogene serine/threonine-protein kinase pim-1
Host Species Rabbit
Immunogen A synthetic peptide of human PIM1 (Uniprot#: P11309)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Oncogenes, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5292
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.