missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM1 Antibody (ST0513), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-67528
This item is not returnable.
View return policy
Description
PIM1 Monoclonal specifically detects PIM1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PIM1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:400, Immunohistochemistry-Frozen | |
| EC 2.7.11.1, Oncogene PIM1, PIM, pim-1 kinase 44 kDa isoform, pim-1 oncogene, pim-1 oncogene (proviral integration site 1), proto-oncogene serine/threonine-protein kinase pim-1 | |
| Rabbit | |
| Protein A purified | |
| Cancer, Oncogenes, Protein Kinase, Tumor Suppressors | |
| 5292 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| ST0513 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| PIM1 | |
| Synthetic peptide within human PIM1 aa 20-60. (SwissProt: P11309 Human; SwissProt: P06803 Mouse) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction