missing translation for 'onlineSavingsMsg'
Learn More

Pim1/AKK1-IN-1, MedChemExpress

Product Code. p-200098673
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493898

Brand: Medchem Express HY10371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pim1/AKK1-IN-1 is a potent multi-kinase inhibitor with Kd values of 35 nM/53 nM/75 nM/380 nM for Pim1/AKK1/MST2/LKB1 respectively, and also inhibits MPSK1 and TNIK.

Pim1/AKK1-IN-1 is a potent multi-kinase inhibitor with Kd values of 35 nM/53 nM/75 nM/380 nM for Pim1/AKK1/MST2/LKB1 respectively, and also inhibits MPSK1 and TNIK.

Specifications

Chemical Name or Material Pim1/AKK1-IN-1
CAS 1093222-27-5
Color Gray
Purity Grade Notes Research
Molecular Formula C20H13N5O
Quantity 10 mg
Synonym LKB1/AAK1 dual inhibitor
Solubility Information DMSO : 50 mg/mL (147.34 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C2=CN=C3C(C(NC(C4=CC=CN=C4)=O)=CN3)=C2)C=C1
Molecular Weight (g/mol) 339.35
Formula Weight 339.35
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.