missing translation for 'onlineSavingsMsg'
Learn More

Pim1/AKK1-IN-1, MedChemExpress

Product Code. 16493898
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493898 10 mg 10mg
16426627 5 mg 5mg
16420267 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493898 Supplier Medchem Express Supplier No. HY10371
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pim1/AKK1-IN-1 is a potent multi-kinase inhibitor with Kd values of 35 nM/53 nM/75 nM/380 nM for Pim1/AKK1/MST2/LKB1 respectively, and also inhibits MPSK1 and TNIK.

Pim1/AKK1-IN-1 is a potent multi-kinase inhibitor with Kd values of 35 nM/53 nM/75 nM/380 nM for Pim1/AKK1/MST2/LKB1 respectively, and also inhibits MPSK1 and TNIK.

Specifications

Chemical Name or Material Pim1/AKK1-IN-1
CAS 1093222-27-5
Color Gray
Purity Grade Notes Research
Molecular Formula C20H13N5O
Quantity 10 mg
Synonym LKB1/AAK1 dual inhibitor
Solubility Information DMSO : 50 mg/mL (147.34 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C2=CN=C3C(C(NC(C4=CC=CN=C4)=O)=CN3)=C2)C=C1
Molecular Weight (g/mol) 339.35
Formula Weight 339.35
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.