missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brusatol, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Brusatol (NSC 172924) is a unique inhibitor of the Nrf2 pathway that sensitizes a broad spectrum of cancer cells to Cisplatin and other chemotherapeutic agents. Brusatol enhances the efficacy of chemotherapy by inhibiting the Nrf2-mediated defense mechanism. Brusatol can be developed into an adjuvant chemotherapeutic agent. Brusatol increases cellular apoptosis.
Specifications
Specifications
| Chemical Name or Material | Brusatol |
| CAS | 14907-98-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H32O11 |
| Quantity | 5 mg |
| Synonym | NSC 172924 |
| Solubility Information | DMSO : 25 mg/mL (48.03 mM; Need ultrasonic) |
| SMILES | CC([C@](C[C@@](O1)2[H])3[H])=C(O)C(C[C@]3(C)[C@]4([H])[C@]2(CO[C@@]5([C@@H](O)[C@@H]4O)C(OC)=O)[C@@]5([H])[C@@H](OC(/C=C(C)/C)=O)C1=O)=O |
| Molecular Weight (g/mol) | 520.53 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction