missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pim1/AKK1-IN-1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Pim1/AKK1-IN-1 is a potent multi-kinase inhibitor with Kd values of 35 nM/53 nM/75 nM/380 nM for Pim1/AKK1/MST2/LKB1 respectively, and also inhibits MPSK1 and TNIK.
Specifications
Specifications
| Chemical Name or Material | Pim1/AKK1-IN-1 |
| CAS | 1093222-27-5 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C20H13N5O |
| Quantity | 50 mg |
| Synonym | LKB1/AAK1 dual inhibitor |
| Solubility Information | DMSO : 50 mg/mL (147.34 mM; Need ultrasonic) |
| SMILES | N#CC1=CC=C(C2=CN=C3C(C(NC(C4=CC=CN=C4)=O)=CN3)=C2)C=C1 |
| Molecular Weight (g/mol) | 339.35 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction