missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILRA, Mouse anti-Human, Clone: 3C2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00029992-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MGRPLLLPLLPLLLPPAFLQPSGSTGSGPSYLYGVTQPKHLSASMGGSVEIPFSFYYPWELATAPDVRISWRRGHFHGQSFYSTRPPSIHKDYVNRLFLNWTEGQKSGFLRISNLQKQDQSVYFCRVELDTRSSGRQQWQSIEGTKLSITQGQQRTKATTPAREPFQNTEEPYENIRNEGQNTDPKLNPKLHLTQSTSQPPSPQEPPERDPVLCLKGLTNGQPSQDSpecifications
| PILRA | |
| Monoclonal | |
| Unconjugated | |
| paired immunoglobin-like type 2 receptor alpha | |
| FDF03 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3C2 | |
| In 1x PBS, pH 7.4 | |
| BC017812 | |
| PILRA | |
| PILRA (AAH17812, 1 a.a. ∼ 226 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 29992 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction