missing translation for 'onlineSavingsMsg'
Learn More

PILR-alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £484.00

Specifications

Antigen PILR-alpha
Dilution Immunohistochemistry-Paraffin 1:500 - 1:1000
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18222745
View Documents
Novus Biologicals
NBP2-55310
100 μL
£484.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18649206
View Documents
Novus Biologicals
NBP2-55310-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PILR-alpha Polyclonal specifically detects PILR-alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PILR-alpha
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha
PILRA
IgG
Affinity Purified
Immunohistochemistry-Paraffin 1:500 - 1:1000
Polyclonal
Rabbit
Signal Transduction
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
29992
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ALSSSTSPRAPPSHRPLKSPQNETLYSVLK
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.