missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILR-alpha Antibody (2175B) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB64842JF669
This item is not returnable.
View return policy
Description
PILR-alpha Monoclonal antibody specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry,Neutralization
Specifications
| PILR-alpha | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha | |
| Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag, Gln20-Thr196, Accession # Q9UKJ1 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Neutralization | |
| 2175B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Signal Transduction | |
| 29992 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction