missing translation for 'onlineSavingsMsg'
Learn More

Pilocarpine hydrochloride, 99.3%, MP Biomedicals™

Product Code. 11447122 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447122 10 g 10g
11427122 1 g 1g
11437122 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447122 Supplier MP Biomedicals Supplier No. 0215189210
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 54-71-7
Molecular Formula C11H17ClN2O2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00012722
InChI Key RNAICSBVACLLGM-GNAZCLTHSA-N
Synonym pilocarpine hydrochloride, +-pilocarpine hydrochloride, pilocarpal, pilocar, pilocar smp, almocarpine, pilomiotin, pilovisc, pilocel, pilocarpine hcl
PubChem CID 5909
IUPAC Name hydrogen (3S,4R)-3-ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-one chloride
SMILES [H+].[Cl-].CC[C@H]1[C@@H](CC2=CN=CN2C)COC1=O

Specifications

Assay Percent Range 0.993
Quantity 10 g
Formula Weight 244.7
Physical Form Crystalline
Chemical Name or Material Pilocarpine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.