missing translation for 'onlineSavingsMsg'
Learn More

Pilaralisib, MedChemExpress

Product Code. 16433117
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16433117

Brand: Medchem Express HY16526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pilaralisib (XL147; SAR245408) is a potent and highly selective class I PI3Ks inhibitor with IC50s of 39 nM, 383 nM, 23 nM and 36 nM for PI3Kα, PI3Kβ, PI3Kγ, and PI3Kδ.

Pilaralisib (XL147

Specifications

Chemical Name or Material Pilaralisib
CAS 934526-89-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H25ClN6O4S
Quantity 50 mg
Synonym XL-147 SAR245408
Solubility Information DMSO : ≥ 100 mg/mL (184.84 mM)
SMILES CC(C)(N)C(NC1=CC=CC(S(=O)(NC2=NC3=CC=CC=C3N=C2NC4=CC(OC)=CC=C4Cl)=O)=C1)=O
Molecular Weight (g/mol) 541.02
Formula Weight 541.02
Percent Purity 95.14%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.