missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plate Frame for 24-well Piko PCR Plates, white

Product Code. 11355960 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11355960 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11355960 Supplier Thermo Scientific™ Supplier No. SFR0241

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.

Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.

Piko PCR Plates are also flexible alternatives for conventional microplates. Four plates, each the size of a microscope slide, can be reversibly snapped into a rigid frame, producing the equivalent of a standard 96- or 384-well microplate. This format allows multiple sample sources to be combined into a single PCR run, or subjected to different post-PCR processing.
  • Wall thickness half that of conventional thin-walled tubes
  • Flexible format saves cost and reduces waste
  • Compatible with standard multi-channel pipettes and liquid handlers
  • Well spacing and footprint correspond to industry (ANSI) dimensions
  • Available in clear and white

Recommended for:

PCR, Fast PCR, High Performance PCR

TRUSTED_SUSTAINABILITY

Specifications

Packaging 50 Frame
Product Line Piko
Color White
Product Type Frame
For Use With (Equipment) Piko Thermal Cycler, PikoReal Real-Time PCR System
Material Polypropylene
No. of Wells 24
Plate Compatibility 24-Well Plates
Quantity 50 frames
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.