missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Piko PCR Plate, 96-well, white

Product Code. 10056769 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200/Cs.
50 Plates
Unit Size:
Pack of 200
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10056769 200/Cs. Pack of 200
15206217 50 Plates Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10056769 Supplier Thermo Scientific™ Supplier No. SPL0961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Speed up PCR and qPCR applications with Piko 24-well PCR Plates, with ultra-thin wall (UTW) technology to dramatically reduce the thickness of the well walls.

Piko PCR Plates are a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.

Piko PCR Plates are also flexible alternatives for conventional microplates. Four plates, each the size of a microscope slide, can be reversibly snapped into a rigid frame, producing the equivalent of a standard 96- or 384-well microplate. This format allows multiple sample sources to be combined into a single PCR run, or subjected to different post-PCR processing.
  • Wall thickness half that of conventional thin-walled tubes
  • Flexible format saves cost and reduces waste
  • Compatible with standard multi-channel pipettes and liquid handlers
  • Well spacing and footprint correspond to industry (ANSI) dimensions
  • Available in clear and white

Recommended for:

PCR, Fast PCR, High Performance PCR

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 200 Plate
Product Line Piko
Color White
Product Type PCR Microplate
For Use With (Equipment) Piko Thermal Cycler, PikoReal Real-Time PCR System
Material Polypropylene
No. of Wells 96
Quantity 200/Cs.
Reaction Speed Standard
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.