Learn More
Thermo Scientific™ Piko PCR Plate, 96-well, clear
Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.
Brand: Thermo Scientific™ SPL096050
54.28 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.50000kg
Descrizione
Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.
Four plates can be snapped into a single frame producing the equivalent of a standard 384-well microplate.Features
• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 96-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 384-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions
Applications
• PCR
• qPCR/Real-Time PCR
Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Plate Frame for 96-well Piko PCR Plates, white
Specifica
No Barcode | |
50 Plate | |
Clear | |
Piko Thermal Cycler, PikoReal Real-Time PCR System | |
96 |
Certified RNase/DNase-free | |
Piko | |
PCR Microplate | |
Polypropylene | |
Standard |