missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIKFYVE Recombinant Rabbit Monoclonal Antibody (HL3158)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556841
This item is not returnable.
View return policy
Description
PIKFYVE Recombinant Monoclonal Antibody for Western Blot
PIKFYVE belongs to a large family of lipid kinases that alter the phosphorylation status of intracellular phosphatidylinositol. The content of phosphatidylinositol 3,5-bisphosphateP2) in endosomal membranes changes dynamically with fission and fusion events that generate or absorb intracellular transport vesicles. PIKFYVE is the PtdIns(3,5)P2-producing component of a trimolecular complex that tightly regulates the level of PtdIns(3,5)P2. Other components of this complex are the PIKFYVE activator VAC14 and the PtdIns(3,5)P2 phosphatase FIG4. In addition to its phosphoinositide 5-kinase activity, PIKFYVE also has protein kinase activity. For a review, see Shisheva.
Specifications
| PIKFYVE | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y2I7 | |
| PIKFYVE | |
| Recombinant fragment of human PIKfyve. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3158 | |
| Unconjugated | |
| PIKFYVE | |
| 1-phosphatidylinositol 3-phosphate 5-kinase; 1-phosphatidylinositol-3-phosphate 5-kinase; 5230400C17Rik; CFD; epididymis luminal protein 37; FAB1; FYVE finger-containing phosphoinositide kinase; HEL37; Kiaa0981; p235; Phosphatidylinositol 3-phosphate 5-kinase; Phosphatidylinositol 3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate 5-kinase type III; phosphatidylinositol-3-phosphate/phosphatidylinositol 5-kinase, type III; phosphatidylinositol-4-phosphate 5-kinase, type III; phosphoinositide kinase, FYVE finger containing; phosphoinositide kinase, FYVE-type zinc finger containing; PIKfyve; Pip5k; PIP5K3; Pipk5k3; PipkIII; PIPkin-III; PtdIns(4)P-5-kinase; type III PIP kinase; ZFYVE29; zinc finger, FYVE domain containing 29 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 200576 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction