missing translation for 'onlineSavingsMsg'
Learn More

PIKFyve/PIP5K3 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30736631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736631 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736631 Supplier Novus Biologicals Supplier No. AF7885JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PIKFyve/PIP5K3 Polyclonal antibody specifically detects PIKFyve/PIP5K3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIKFyve/PIP5K3
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias 1-phosphatidylinositol-3-phosphate 5-kinase, EC 2.7.1.150, FLJ37746, FYVE finger-containing phosphoinositide kinase, KIAA0981CFD, MGC40423, p235, Phosphatidylinositol-3-phosphate 5-kinase, Phosphatidylinositol-3-phosphate 5-kinase type III, phosphoinositide kinase, FYVE finger containing, PIKfyve, PIP5K, PIP5K3FAB1, PIPkin-III, type III, Type III PIP kinase
Host Species Sheep
Immunogen E. coli-derived recombinant human PIKFyve, Lys215-Lys361, Accession # Q9Y2I7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 200576
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.