missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pikachurin Mouse anti-Human, Clone: 1C6D12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Pikachurin |
|---|---|
| Clone | 1C6D12 |
| Concentration | 2 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807925
|
Proteintech
66232-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897915
|
Proteintech
66232-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This monoclonal antibody detects two bands of 111 kDa and 55-60 kDa. The lower-molecular-weight band has been reported and likely represents a fragment resulting from cleavage of Pikachurin.
Involved in both the retinal photoreceptor ribbon synapse formation and physiological functions of visual perception. Necessary for proper bipolar dendritic tip apposition to the photoreceptor ribbon synapse. Promotes matrix assembly and cell adhesiveness.Specifications
| Pikachurin | |
| 2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Agrin like protein, AGRINL, AGRNL, EGFLAM, Pikachurin | |
| EGFLAM | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1C6D12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q63HQ2 | |
| 133584 | |
| Pikachurin Fusion Protein Ag6362 | |
| Primary | |
| -20°C | |
| EGFLAM |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title