missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R6, Mouse anti-Human, Clone: 1G16, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00146850-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: GKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQSpecifications
| PIK3R6 | |
| Monoclonal | |
| Unconjugated | |
| phosphoinositide-3-kinase, regulatory subunit 6 | |
| C17orf38/DKFZp666P158/FLJ34500/HsT41028/p84/p87(PIKAP)/p87PIKAP | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1G16 | |
| In 1x PBS, pH 7.4 | |
| NM_001010855 | |
| PIK3R6 | |
| PIK3R6 (NP_001010855, 661 a.a. ∼ 754 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 146850 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction