missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48560
This item is not returnable.
View return policy
Description
PIK3R6 Polyclonal antibody specifically detects PIK3R6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PIK3R6 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| C17orf38, chromosome 17 open reading frame 38, FLJ34500, HsT41028, p84, p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein, p87 PI3K adapter protein, p87(PIKAP), p87PIKAPDKFZp666P158, Phosphoinositide 3-kinase gamma adapter protein of 87 kDa, phosphoinositide 3-kinase regulatory subunit 6, phosphoinositide-3-kinase, regulatory subunit 6, PI3Kgamma adapter protein of 87 kDa | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PAAPVTDHTCLNVNVTEVVKSSNLAGKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKP | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 146850 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction