missing translation for 'onlineSavingsMsg'
Learn More

PIK3R5 Antibody (OTI4G9), DyLight 594, Novus Biologicals™

Product Code. 18411157 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411157 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411157 Supplier Novus Biologicals Supplier No. NBP273408DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIK3R5 Monoclonal antibody specifically detects PIK3R5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3R5
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4G9
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias F730038I15Rik, FOAP-2, p101, P101-PI3K, Phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit, phosphoinositide-3-kinase, regulatory subunit 5, phosphoinositide-3-kinase, regulatory subunit, polypeptide p101, PI3-kinase p101 subunit, PI3-kinase regulatory subunit 5, PtdIns-3-kinase regulatory subunit, regulatory subunit 5, p101
Host Species Mouse
Immunogen Full length recombinant protein of human PIK3R5 (NP_055123) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23533
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.