missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3R4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583302
This item is not returnable.
View return policy
Description
Immunogen sequence: GGRVKTLTFC QGSHYLAIAS DNGAVQLLGI EASKLPKSPK IHPLQSRILD QKEDGCVVDM HHFNSGAQSV LAYATVNGSL VGWDLRSSSN AWTL Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Regulatory subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate; different complex forms are believed to play a role in multiple membrane trafficking pathways: PI3KC3-C1 is involved in initiation of autophagosomes and PI3KC3-C2 in maturation of autophagosomes and endocytosis. Involved in regulation of degradative endocytic trafficking and cytokinesis, probably in the context of PI3KC3-C2 (PubMed:20643123). [UniProt].
Specifications
| PIK3R4 | |
| Polyclonal | |
| Unconjugated | |
| PIK3R4 | |
| 2210010O15Rik; C730038E05Rik; C85833; D9Ertd418e; p150; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 4, p150; phosphatidylinositol 3-kinase-associated p150; phosphoinositide 3-kinase adaptor protein; Phosphoinositide 3-kinase regulatory subunit 4; phosphoinositide-3-kinase regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4; phosphoinositide-3-kinase, regulatory subunit 4, p150; PI3-Kinase p150; PI3-kinase p150 subunit; PI3-kinase regulatory subunit 4; PI3-kinase VPS15; PIK3R4; VPS15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99570 | |
| PIK3R4 | |
| Recombinant protein corresponding to Human PIK3R4. Recombinant protein control fragment (Product #RP-97253). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction