missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R4, Mouse anti-Human, Clone: 1H7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00030849-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: AGSDMKIRFWDLAYPERSYVVAGSTSSPSVSYYRKIIEGTEVVQEIQNKQKVGPSDDTPRRGPESLPVGHHDIITDVATFQTTQGFIVTASRDGIVKVWKSpecifications
| PIK3R4 | |
| Monoclonal | |
| Unconjugated | |
| phosphoinositide-3-kinase, regulatory subunit 4 | |
| MGC102700/VPS15/p150 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 1H7 | |
| In 1x PBS, pH 7.4 | |
| NM_014602 | |
| PIK3R4 | |
| PIK3R4 (NP_055417, 1259 a.a. ∼ 1358 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 30849 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction