missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3R4 Antibody (SR1197), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | PIK3R4 |
|---|---|
| Clone | SR1197 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647084
|
Novus Biologicals
NBP3-22308-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652275
|
Novus Biologicals
NBP3-22308-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIK3R4 Monoclonal antibody specifically detects PIK3R4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| PIK3R4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11.1, MGC102700, p150, phosphatidylinositol 3-kinase-associated p150, Phosphoinositide 3-kinase adaptor protein, phosphoinositide 3-kinase regulatory subunit 4, phosphoinositide-3-kinase, regulatory subunit 4, phosphoinositide-3-kinase, regulatory subunit 4, p150, PI3-kinase p150 subunit, PI3-kinase regulatory subunit 4, VPS 15, VPS15 | |
| A synthesized peptide derived from human PIK3R4 (Uniprot #: Q99570) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1197 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 30849 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title