missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIK3R4 Polyclonal antibody specifically detects PIK3R4 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PIK3R4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.11.1, MGC102700, p150, phosphatidylinositol 3-kinase-associated p150, Phosphoinositide 3-kinase adaptor protein, phosphoinositide 3-kinase regulatory subunit 4, phosphoinositide-3-kinase, regulatory subunit 4, phosphoinositide-3-kinase, regulatory subunit 4, p150, PI3-kinase p150 subunit, PI3-kinase regulatory subunit 4, VPS 15, VPS15 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 1075 and 1125 of human vacuolar protein sorting 15 using the numbering given in entry NP_055417.1 (GeneID 30849). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?