missing translation for 'onlineSavingsMsg'
Learn More

PIK3R4 Antibody (1B5), Novus Biologicals™

Product Code. 18368089 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368089 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368089 Supplier Novus Biologicals Supplier No. H00030849M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIK3R4 Monoclonal antibody specifically detects PIK3R4 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3R4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1B5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055417
Gene Alias EC 2.7.11.1, MGC102700, p150, phosphatidylinositol 3-kinase-associated p150, Phosphoinositide 3-kinase adaptor protein, phosphoinositide 3-kinase regulatory subunit 4, phosphoinositide-3-kinase, regulatory subunit 4, phosphoinositide-3-kinase, regulatory subunit 4, p150, PI3-kinase p150 subunit, PI3-kinase regulatory subunit 4, VPS 15, VPS15
Host Species Mouse
Immunogen PIK3R4 (NP_055417, 1259 a.a. ~ 1358 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AGSDMKIRFWDLAYPERSYVVAGSTSSPSVSYYRKIIEGTEVVQEIQNKQKVGPSDDTPRRGPESLPVGHHDIITDVATFQTTQGFIVTASRDGIVKVWK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 30849
Target Species Human, Mouse, Monkey
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.