missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3IP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16826-1-AP-150UL
This item is not returnable.
View return policy
Description
The exact function of PIK3IP1 remains unknown.Specifications
| PIK3IP1 | |
| Polyclonal | |
| Unconjugated | |
| PIK3IP1 | |
| HGFL, hHGFL(S), PIK3IP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 113791, 216505, 305472 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q56A20, Q7TMJ8, Q96FE7 | |
| Pik3ip1 | |
| PIK3IP1 Fusion Protein Ag10427 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction