missing translation for 'onlineSavingsMsg'
Learn More

PIK3IP1 Antibody [FITC], Novus Biologicals™

Product Code. 30761980 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30761980 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30761980 Supplier Novus Biologicals Supplier No. NBP347142F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIK3IP1 Polyclonal antibody specifically detects PIK3IP1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3IP1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias HGFLhHGFL(S), Kringle domain-containing protein HGFL, MGC17330, phosphoinositide-3-kinase interacting protein 1, phosphoinositide-3-kinase-interacting protein 1
Host Species Rabbit
Immunogen phosphoinositide-3-kinase interacting protein 1. (Uniprot# Q96FE7)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113791
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.