missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIK3IP1 Polyclonal antibody specifically detects PIK3IP1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | PIK3IP1 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HGFLhHGFL(S), Kringle domain-containing protein HGFL, MGC17330, phosphoinositide-3-kinase interacting protein 1, phosphoinositide-3-kinase-interacting protein 1 |
| Host Species | Rabbit |
| Immunogen | phosphoinositide-3-kinase interacting protein 1. (Uniprot# Q96FE7) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?