Learn More
Invitrogen™ Human PIK3CB/PIK3R1 (p110 beta/p85 alpha), His Tag Recombinant Protein
Shop All Thermo Scientific ProductsDescription
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. PIK3CB/PIK3R1 (p110β/p85α) Recombinant Human Protein is: Available in 3 convenient sizes Validated in our Adapta™ and Kinase Binding assays A full-length protein that can help advance your research when used in validated assays Related assay platforms:LanthaScreen™ Kinase Binding AssayAdapta™ Kinase Activity Assay Protein Form: Full Length, Recombinant, Active The gene accession number for PIK3CB (p110β) is NP_006210 and for PIK3R1 (p85α) is NP_852664.1.
Specifications
Specifications
| Accession Number | P27986, P42338 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM sodium phosphate with 150mM Imidazole, 25% glycerol, 0.1mM PMSF, 300mM NaCl, 0.25mM DTT and no preservative; pH 7 |
| Gene ID (Entrez) | 5291, 5295 |
| Molecular Weight (g/mol) | 123.7-84.7 kDa |
| Name | Human PIK3CB/PIK3R1 (p110 beta/p85 alpha), His Tag |
| pH Range | 7 |
| Purification Method | Purified |
| Quantity | 20 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.