missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3CB, Mouse anti-Human, Clone: 10D5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005291-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: EFRRKMRKFSEEKILSLVGLSWMDWLKQTYPPEHEPSIPENLEDKLYGGKLIVAVHFENCQDVFSFQVSPNMNPIKVNELAIQKRLTIHGKEDEVSPYDYVLQVSGRVEYSpecifications
| PIK3CB | |
| Monoclonal | |
| Unconjugated | |
| phosphoinositide-3-kinase, catalytic, beta polypeptide | |
| DKFZp779K1237/MGC133043/PI3K/PI3KCB/PI3Kbeta/PIK3C1/p110-BETA | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, In situ PLA, Western Blot | |
| 10D5 | |
| In 1x PBS, pH 7.4 | |
| NM_006219 | |
| PIK3CB | |
| PIK3CB (NP_006210, 147 a.a. ∼ 256 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5291 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction