Learn More
Invitrogen™ Human PIK3CA/PIK3R3 (p110 alpha/p55 gamma), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3CA/PIK3R3 (p110 alpha /p55 gamma) recombinant human protein (full length) was expressed in insect cells. PIK3CA/PIK3R3 (p110 alpha /p55 gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIK3CA is NP_006209.2 and for PIK3R3 is AAH21622.
Specifications
Specifications
| Accession Number | P42336, Q92569 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM sodium phosphate with 150mM Imidazole, 0.25mM DTT, 25% glycerol, 0.1mM PMSF, 300mM NaCl and no preservative; pH 7 |
| Gene ID (Entrez) | 5290, 8503 |
| Molecular Weight (g/mol) | 125.4-56.3 kDa |
| Name | Human PIK3CA/PIK3R3 (p110 alpha/p55 gamma), His Tag |
| pH Range | 7 |
| Purification Method | Purified |
| Quantity | 20 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.