missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIK3CA/PIK3R3 (p110 alpha/p55 gamma), His Tag Recombinant Protein

Product Code. 15980313
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
This item is not returnable. View return policy

Product Code. 15980313

Brand: Invitrogen™ A31082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3CA/PIK3R3 (p110 alpha /p55 gamma) recombinant human protein (full length) was expressed in insect cells. PIK3CA/PIK3R3 (p110 alpha /p55 gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIK3CA is NP_006209.2 and for PIK3R3 is AAH21622.

PIK3CA/PIK3R3 lipid kinase (p110 alpha /p55 gamma) or Phosphatidylinositol 3-kinase (PIK3) phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position which serves as second messenger in growth signaling pathways. PIK3CA comprises a 110 kDa catalytic subunit and a regulatory subunit of either 85, 55, or 50 kDa. PIK3CA plays an important role in the metabolic actions of insulin, and a mutation in the PIK3CA has been associated with insulin resistance and plays an important role in glucose homeostasis in vivo. PIK3CA also plays an essential role in the development and induction of mast cells in normal and pathogenic immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42336, Q92569
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM sodium phosphate with 150mM Imidazole, 0.25mM DTT, 25% glycerol, 0.1mM PMSF, 300mM NaCl and no preservative; pH 7
Gene ID (Entrez) 5290, 8503
Molecular Weight (g/mol) 125.4-56.3 kDa
Name Human PIK3CA/PIK3R3 (p110 alpha/p55 gamma), His Tag
pH Range 7
Purification Method Purified
Quantity 20 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias PIK3CA; PIK3R3
Common Name PIK3CA/PIK3R3
Gene Symbol PIK3CA, PIK3R3
KinaseFamily PI3 Kinase
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.