missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIK3CA E542K/PIK3R1 (p110 alpha E542K/p85 alpha), His Tag Recombinant Protein

Product Code. p-7181649
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
This item is not returnable. View return policy

Product Code. 15910423

Brand: Invitrogen™ A33518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3CA E542K/PIK3R1 (p110 alpha E542K/p85 alpha) recombinant human protein (full length) was expressed in insect cells. PIK3CA E542K/PIK3R1 (p110 alpha E542K/p85 alpha) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active, Mutant The gene accession number for PIK3CA (p110α) is NP_006209.2 and for PIK3R1 (p85α) is NP_852664.1.

PIK3CA E542K/PIK3R1 (p110 alpha E542K/p85 alpha) lipid kinase is comprised of a 110 kDa catalytic subunit and an 85 kDa regulatory subunit. A number of isoforms of the 110 kDa catalytic subunit and the 85 kDa regulatory subunit exist in cells. PIK3CA is frequently mutated or amplified in a variety of cancers including ovarian and colon and this kinase is one of the PI3K mutants. PIK3CA gene copy number is increased in over 30% of ovarian cancers and this leads to increased PI3-kinase activity. Furthermore, the activity of PIK3CA is essential for vascular development and inactivation of PIK3CA leads to severe defects in angiogenic sprouting and vascular remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27986, P42336
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM sodium phosphate with 300mM NaCl, 0.1mM PMSF, 0.25mM DTT, 150mM Imidazole, 25% glycerol and no preservative; pH 7
Gene ID (Entrez) 5290, 5295
Molecular Weight (g/mol) 125.4-84.7 kDa
Name Human PIK3CA E542K/PIK3R1 (p110 alpha E542K/p85 alpha), His Tag
pH Range 7
Purification Method Purified
Quantity 20 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias PIK3CA; PIK3R1
Common Name PIK3CA (E545K Mutant)/PIK3R1
Gene Symbol PIK3CA, PIK3R1
KinaseFamily PI3 Kinase
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active, Mutant
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.