missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3CA Antibody (10E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26384-50ul
This item is not returnable.
View return policy
Description
PIK3CA Monoclonal antibody specifically detects PIK3CA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry
Specifications
| PIK3CA | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 10E5 | |
| Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| EC 2.7.1, EC 2.7.1.153, MGC142161, MGC142163, p110-alpha, Phosphatidylinositol 3-Kinase catalytic subunit alpha, phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha, phosphatidylinositol 3-kinase, catalytic, alpha polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform, phosphoinositide-3-kinase, catalytic, alpha polypeptide, PI3K, PI3K-alpha, PI3-kinase p110 subunit alpha, PI3-kinase subunit alpha, PtdIns-3-kinase p110, PtdIns-3-kinase subunit alpha, PtdIns-3-kinase subunit p110-alpha | |
| A synthesized peptide derived from Human PIK3CA [UniProt P42336] | |
| 50 μL | |
| Breast Cancer, Cancer, Diabetes Research, mTOR Pathway, Oncogenes | |
| 5290 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction