missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIK3C2G (PI3K-C2 gamma), GST Tag Recombinant Protein

Product Code. 17158805
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
This item is not returnable. View return policy

Product Code. 17158805

Brand: Invitrogen™ A33392

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3C2G (PI3K-C2 gamma) recombinant human protein (468-1203) was expressed in insect cells. PIK3C2G (PI3K-C2 gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for PIK3C2G (PI3K-C2 gamma) is AAI30278.1.

PIK3C2G lipid kinase belongs to the phosphoinositide 3-kinase (PI3K) family which play major roles in signaling pathways and are involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. PIK3C2G contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases that act as calcium-dependent phospholipid binding motifs which mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The N-terminal region of PIK3C2G and the C-terminal C2 domain are required for enzymatic activity. The PI 3-kinase C2gamma gene encodes a 1,050 amino acid polypeptide with 36% identity to human PI 3-kinase C2alpha. Research indicates that PI 3-kinase C2gamma can block the growth of human colon cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75747
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.1mM PMSF, 0.25mM DTT, 25% glycerol, 0.1mM EDTA, 10mM glutathione, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 5288
Molecular Weight (g/mol) 110 kDa
Name Human PIK3C2G (PI3K-C2 gamma), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C80387; phosphatidylinositol 3-kinase, C2 domain containing, gamma polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit gamma; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing gamma polypeptide; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit gamma; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 gamma; phosphoinositide 3-Kinase-C2-gamma; phosphoinositide-3-kinase, class 2, gamma polypeptide; PI 3-kinase C2gamma; PI3KC2gamma; PI3K-C2gamma; PI3K-C2-gamma; Pik3c2g; PTDINS-3-kinase C2 gamma; ptdIns-3-kinase C2 subunit gamma
Common Name PIK3C2G
Gene Symbol PIK3C2G
KinaseFamily PI3 Kinase
Protein Length 468-1203
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 468-1203
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.