missing translation for 'onlineSavingsMsg'
Learn More

PIK3C2G Antibody [Biotin], Novus Biologicals™

Product Code. 30762595 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762595 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762595 Supplier Novus Biologicals Supplier No. NBP347143B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIK3C2G Polyclonal antibody specifically detects PIK3C2G in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIK3C2G
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias EC 2.7.1, EC 2.7.1.154, MGC163149, phosphatidylinositol 3-kinase C2 domain-containing gamma, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing gammapolypeptide, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit gamma, Phosphoinositide 3-kinase-C2-gamma, phosphoinositide-3-kinase, class 2, gamma polypeptide, PI3K-C2-gamma, PI3K-C2GAMMA, PTDINS-3-kinase C2 gamma, PtdIns-3-kinase C2 subunit gamma
Host Species Rabbit
Immunogen phosphoinositide-3-kinase, class 2, gamma polypeptide. (Uniprot# O75747)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5288
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.