missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3C2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100895
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PIK3C2B.
The Class II PI3 kinases, including PIK3C2B, phosphorylate the 3 position of PI or PI4P. Though not as well studied as the class I PI3 kinases, the class II enyzmes have been linked to diverse, receptor-mediated cellular processes including insulin signaling, neuronal survival, and growth factor signaling. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
Specifications
| PIK3C2B | |
| Polyclonal | |
| Unconjugated | |
| PIK3C2B | |
| C2-PI3K; C330011J12Rik; Gm199; phosphatidylinositol 3-kinase C2 domain-containing beta polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit beta; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 beta; phosphoinositide 3-kinase-C2-beta; phosphoinositide-3-kinase, class 2, beta polypeptide; pi3kc2b; PI3KC2beta; PI3K-C2beta; PI3K-C2-beta; PIK3C2B; PTDINS-3-kinase C2 beta; ptdIns-3-kinase C2 subunit beta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 240752, 5287 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00750 | |
| PIK3C2B | |
| A synthesized peptide derived from human PIK3C2B(Accession O00750), corresponding to amino acid residues F790-W840. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction