missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3C2A Monoclonal Antibody (OTI3D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526509
This item is not returnable.
View return policy
Description
PIK3C2A Monoclonal Antibody for Western Blot, IHC (P)
The Class II PI3 kinases, including PIK3C2A, phosphorylate the 3 position of PI or PI4P. Though not as well studied as the Class I PI3 kinases, the Class II enyzmes have been linked to diverse, receptor-mediated cellular processes including insulin signaling, neuronal survival, and growth factor signaling. PI3-kinases play roles in signaling pathways involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. This protein contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases. C2 domains act as calcium-dependent phospholipid binding motifs that mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The PI3-kinase activity of this protein is not sensitive to nanomolar levels of the inhibitor wortmanin. This protein was shown to be able to be activated by insulin and may be involved in integrin-dependent signaling.
Specifications
| PIK3C2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00443 | |
| PIK3C2A | |
| Human recombinant protein fragment corresponding to amino acids 230-560 of PIK3C2A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D9 | |
| Unconjugated | |
| PIK3C2A | |
| C2-containing phosphatidylinositol kinase; CPK; Cpk-m; DKFZp686L193; MGC142218; p170; phosphatidylinositol 3-kinase, C2 domain containing, alpha polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha; phosphoinositide 3-kinase-C2-alpha; phosphoinositide-3-kinase, class 2, alpha polypeptide; PI3KC2; PI3-K-C2(ALPHA); pi3kc2a; PI3-K-C2A; PI3K-C2alpha; PI3K-C2-alpha; Pik3c2a; ptdIns-3-kinase C2 subunit alpha | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5286 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction