missing translation for 'onlineSavingsMsg'
Learn More

PIK-294, MedChemExpress

Product Code. 16464836
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464836 100 mg 100mg
16446507 5 mg 5mg
16403788 10 mg 10mg
16400167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464836 Supplier Medchem Express Supplier No. HY10303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PIK-294 is a potent p110δ-selective inhibitor with an IC50 of 10 nM.

PIK-294 is a potent p110δ-selective inhibitor with an IC50 of 10 nM.

Specifications

Chemical Name or Material PIK-294
CAS 900185-02-6
Color White
Purity Grade Notes Research
Molecular Formula C28H23N7O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (204.28 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=CC=C2C)C(CN3N=C(C(C3=NC=N4)=C4N)C5=CC(O)=CC=C5)=NC6=CC=CC(C)=C16
Molecular Weight (g/mol) 489.53
Formula Weight 489.53
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.