missing translation for 'onlineSavingsMsg'
Learn More

PIH1D1 Antibody (1E8), Novus Biologicals™

Product Code. 18383909 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383909 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383909 Supplier Novus Biologicals Supplier No. H00055011M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIH1D1 Monoclonal antibody specifically detects PIH1D1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIH1D1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01108
Gene Alias NOP17FLJ20643, Nucleolar protein 17 homolog, PIH1 domain containing 1, PIH1 domain-containing protein 1
Host Species Mouse
Immunogen FLJ20643 (AAH01108, 1 a.a. ~ 290 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MANPKLLGLELSEAEAIGADSARFEELLLQASKELQQAQTTRPESTQIQPQPGFCIKTNSSEGKVFINICHSPSIPPPADVTEEELLQMLEEDQAGFRIPMSLGEPHAELDAKGQGCTAYDVAVNSDFYRRMQNSDFLRELVITIAREGLEDKYNLQLNPEWRMMKNRPFMGSISQQNIRSEQRPRIQELGDLYTPAPGRAESGPEKPHLNLWLEAPDLLLAEIDLPKLDGALGLSLEIGENRLVMGGPQQLYHLDAYIPLQINSHESKAAFHRKRKQLMVAMPLLLVPS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 55011
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.