missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGY Polyclonal Antibody

Description
Despite its predicted molecular weight, PIG-Y often migrates at 28-30kDa in SDS-PAGE. A suggested positive control is A-20 cell lysate. PA5-20651 can be used with blocking peptide PEP-0771.
Glycosylphosphatidylinositol (GPI) lipid anchoring is an important post-translational modification of proteins that takes place in the endoplasmic reticulum. The synthesis of GPI is initiated by GPI-N-acetylglucosaminyltransferase (GPI-GnT), a complex of proteins including PIG-A, PIG-H, PIG-C, GPI1, and DPM2. PIG-Y, the mammalian homolog to yeast Eri1p, is also thought to be involved in the biosynthesis of GPI. The PIG-Y gene encodes two proteins, one of which arises from leaky scanning of the mRNA.
Specifications
Specifications
| Antigen | PIGY |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PIGY |
| Gene Accession No. | P0C1P0, Q3MUY2 |
| Gene Alias | 1810008A14Rik; HPMRS6; phosphatidylinositol glycan anchor biosynthesis class Y; phosphatidylinositol glycan anchor biosynthesis, class Y-like; phosphatidylinositol N-acetylglucosaminyltransferase subunit Y; Phosphatidylinositol-glycan biosynthesis class Y protein; Pigy; PIG-Y; Pigyl; PREY; protein preY |
| Gene Symbols | PIGY, Pigyl |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?