missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101749
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PIGX.
Essential component of glycosylphosphatidylinositol-mannosyltransferase 1 which transfers the first of the 4 mannoses in the GPI-anchor precursors during GPI-anchor biosynthesis. Probably acts by stabilizing the mannosyltransferase PIGM.
Specifications
| PIGX | |
| Polyclonal | |
| Unconjugated | |
| PIGX | |
| 2010319C14Rik; GPI-mannosyltransferase subunit; phosphatidylinositol glycan anchor biosynthesis class X; phosphatidylinositol glycan anchor biosynthesis, class X; phosphatidylinositol glycan, class X; phosphatidylinositol-glycan biosynthesis class X protein; PIGX; PIG-X | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54965 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TBF5 | |
| PIGX | |
| A synthesized peptide derived from human PIGX(Accession Q8TBF5), corresponding to amino acid residues K183-S233. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction