missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGT Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | PIGT |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PIGT Polyclonal antibody specifically detects PIGT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PIGT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ41596, GPI transamidase component PIG-T, GPI transamidase subunit, MGC8909, NDAP, neurotrophin-regulated neuronal development-associated protein, phosphatidyl inositol glycan class T, phosphatidylinositol glycan anchor biosynthesis, class T, phosphatidylinositol glycan, class T, Phosphatidylinositol-glycan biosynthesis class T protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 260-460 of human PIGT (NP_057021.2). LFRMFSRTLTEPCPLASESRVYVDITTYNQDNETLEVHPPPTTTYQDVILGTRKTYAIYDLLDTAMINNSRNLNIQLKWKRPPENEAPPVPFLHAQRYVSGYGLQKGELSTLLYNTHPYRAFPVLLLDTVPWYLRLYVHTLTITSKGKENKPSYIHYQPAQDRLQPHLLEMLIQLPANSVTKVSIQFERALLKWTEYTPDP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 51604 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title