missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGQ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566389
This item is not returnable.
View return policy
Description
Immunogen sequence: YVYGARLYCLK IHGLSSLWRL FRGKKWNVLR QRVDSCSYDL DQ Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
This gene is involved in the first step in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This gene encodes a N-acetylglucosaminyl transferase component that is part of the complex that catalyzes transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc to phosphatidylinositol (PI).
Specifications
| PIGQ | |
| Polyclonal | |
| Unconjugated | |
| PIGQ | |
| c407A10.1; c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)); glycosylphosphatidylinositol 1 homolog; GPI1; Gpi1h; Gpi1p; Gpih; Mgpi1; MGpi1p; N-acetylglucosaminyl transferase component Gpi1; N-acetylglucosamyl transferase component GPI1; phosphatidylinositol glycan anchor biosynthesis class Q; phosphatidylinositol glycan anchor biosynthesis, class Q; phosphatidylinositol glycan, class Q; phosphatidylinositol N-acetylglucosaminyltransferase subunit Q; Phosphatidylinositol-glycan biosynthesis class Q protein; Pigq; PIG-Q | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9091 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRB3 | |
| PIGQ | |
| Recombinant Human PIGQ. Recombinant protein control fragment (Product #RP-107062). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction