Learn More
PIGP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555464
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NMMSTSPLDS IHTITDNYAK NQQQKKYQEE AIPALRDISI SEVNQMFFLA AKELYTK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
Part of the complex catalyzing the transfer of N- acetylglucosamine from UDP-N-acetylglucosamine to phosphatidylinositol, the first step of GPI biosynthesis.Specifications
PIGP | |
Polyclonal | |
Unconjugated | |
PIGP | |
DCRC; DCRC-S; Down syndrome critical region gene 5; Down syndrome critical region gene c; Down syndrome critical region homolog 5; Down syndrome critical region protein 5; Down syndrome critical region protein 5 homolog; Down syndrome critical region protein C; Dscr5; DSCRC; DSRC; NPD010; phosphatidylinositol glycan anchor biosynthesis class P; phosphatidylinositol glycan anchor biosynthesis, class P; phosphatidylinositol glycan, class P; phosphatidylinositol N-acetylglucosaminyltransferase subunit P; Phosphatidylinositol-glycan biosynthesis class P protein; phosphatidylinositol-n-acetylglucosaminyltranferase subunit; PIGP; PIG-P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51227 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57054 | |
PIGP | |
Recombinant protein corresponding to Human PIGP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only