missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIGP Polyclonal antibody specifically detects PIGP in Human,Mouse,Rat samples. It is validated for ELISA,Serum
Specifications
Specifications
| Antigen | PIGP |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | class P, DCRCDown syndrome critical region protein C, Down syndrome critical region protein 5, DSCR5DCRC-S, DSCRC, DSRC, EC 2.4.1.198, phosphatidylinositol glycan anchor biosynthesis, class P, Phosphatidylinositol-glycan biosynthesis class P protein, phosphatidylinositol-n-acetylglucosaminyltranferase subunit, PIG-P |
| Host Species | Rabbit |
| Immunogen | phosphatidylinositol glycan anchor biosynthesis, class P. (Uniprot# P57054) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?