missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94792-0.1ml
This item is not returnable.
View return policy
Description
PIGP Polyclonal antibody specifically detects PIGP in Mouse samples. It is validated for Western Blot
Specifications
| PIGP | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| class P, DCRCDown syndrome critical region protein C, Down syndrome critical region protein 5, DSCR5DCRC-S, DSCRC, DSRC, EC 2.4.1.198, phosphatidylinositol glycan anchor biosynthesis, class P, Phosphatidylinositol-glycan biosynthesis class P protein, phosphatidylinositol-n-acetylglucosaminyltranferase subunit, PIG-P | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-158 of human PIGP (NP_710148.1). MVENSPSPLPERAIYGFVLFLSSQFGFILYLVWAFIPESWLNSLGLTYWPQKYWAVALPVYLLIAIVIGYVLLFGINMMSTSPLDSIHTITDNYAKNQQQKKYQEEAIPALRDISISEVNQMFFLAAKELYTKN | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51227 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction