missing translation for 'onlineSavingsMsg'
Learn More

PIGO Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18300864 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18300864 20 μg 20µL
18361377 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18300864 Supplier Novus Biologicals Supplier No. NBP31558720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIGO Polyclonal antibody specifically detects PIGO in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGO
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias class O, DKFZp434M222, EC 2.-, FLJ00135, GPI ethanolamine phosphate transferase 3, MGC20536, MGC3079, phosphatidylinositol glycan anchor biosynthesis, class O, Phosphatidylinositol-glycan biosynthesis class O protein, PIG-O, RP11-182N22.4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 950-1050 of human PIGO (NP_116023.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84720
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.