missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGF Antibody (RM0010-8F09), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-85468PE
This item is not returnable.
View return policy
Description
PIGF Monoclonal antibody specifically detects PIGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| PIGF | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| RM0010-8F09 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| GPI11 homolog, MGC32646, MGC33136, phosphatidylinositol glycan anchor biosynthesis, class F, phosphatidylinositol glycan, class F, phosphatidylinositol-glycan biosynthesis class F protein, PIG-F | |
| Mouse recombinant PlGF protein. | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 5281 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction